This product is now discontinued, View Related Products
Oakton® InfraPro Infrared Thermometers 43115
Single laser spot targets the measurement area
  • Measure hazardous targets from a distance
  • InfraPro™ 3 and 4 datalog readings and feature a thermocouple input to take contact temperatures
  • InfraPro™ 5 is rated intrinsically safe, Class I Division 1, Groups A, B, C, D and Class I, Zone 0, AEx ia IIC, T4 at 50°C
Rugged enough for industrial use yet compact and lightweight enough to be carried around with you, these infrared thermometers are extremely easy to use. Just aim, pull the trigger, and read the display. The 3-1/2-digit backlit display shows temperature readings in °F or °C; low-battery indication; and hold, scan, and max. Display holds for 7 seconds. Underrange and overrange conditions are also indicated on the display.
Extra bright laser can be easily seen in bright indoor or outdoor conditions! Advanced models 35639-20, -30, and -40 indicate max, min, dif, avg, emissivity, high alarm, low alarm, probe, log, and recall. Other features include an RTD input, 12 datalogging points, and adjustable emissivity.
All thermometers come with a hard carrying case and feature
6 PRODUCT OPTIONS
Compareitem specifications
Distance To Target Size Ratio (D:S) Emissivity Max Temperature (° F)
Availability Pricing
COMPARE
12:1 Fixed 999
Call For Price
COMPARE
Call For Price
COMPARE
12:1 Fixed 999
Call For Price
COMPARE
30:1 Adjustable 1100
Call For Price
COMPARE
50:1 Adjustable 1400
Call For Price
COMPARE
50:1 Adjustable 1400