Cole Parmer offers a wide range of laboratory incubators for a variety of applications including incubation of micro-organisms and microbiological heating and conditioning. The laboratory incubators available cover temperature ranges from -10oC to 100oC and capacities from 20 litres to 720 litres.
View our selection from leading manufacturers of laboratory incubators or call our application specialists for free support to help you find the right laboratory incubator to best suit your needs.
Natural Convection Laboratory Incubators
Ideal for incubating of micro-organisms, and microbiological heating and conditioning
|
|
Binder Natural Convection Laboratory Incubators
- Capacities from 20 - 720 litres
- Temperatures from ambient +5 to 100oC
- Controller features integrated timer settable from 0 to 99 hours and one adjustable temperature ramp
- Stainless steel interior and coated steel exterior construction
|
|
|
Carbolite Natural Convection Laboratory Incubators
- Capacities from 27 - 215 litres
- Temperatures from ambient +10 to 80oC
- Incubator features a three segment PID microprocessor controller with an adjustable single ramp rate to set point
- Available with a variety of alternate control options
- Up to 15 shelves positions available
|
|
Forced Convection Laboratory Incubators
Ideal for applications with high specimen volumes, such as sample bags and frequent batch changes
Forced models features a high-performance fan to circulate air within the chamber to maximize temperature uniformity and improve recovery times
|
|
Binder Forced Convection Laboratory Incubators
- Capacities from 53 - 720 litres
- Temperatures from +5 to 100oC
- Specially designed air turbines with adjustable airflow rates
- Fast recovering times - 1 to 2 minutes at 37oC
- Controller timer with delayed on, delayed off and temperature dependent off functions
|
|
Carbolite Forced Convection Laboratory Incubators
- Capacities from 28 - 230 litres
- Temperatures from ambient +10 to 80oC
- Designed for long term accuracy and flexibility
- Accuracy of 5oC
|
Refrigerated Convection Laboratory Incubators
Precision incubation control for maintaining specimen quality at low temperatures |
|
|
Binder Refridgerated Laboratory Incubators
- Capacities from 20- 720 litres
- Temperatures from 10oC to 100oC
- Rapid temperature recovery rates: under two minutes at 37oC
- Controller features delayed on, delayed off and temperature dependent delayed off timer functions plus storage capability up to 20 segments
- Hermetically sealed inner glass doors to allow you to monitor samples without influencing chamber conditions
|
|
Carbolite Refridgerated Laboratory Incubators
- Capacities from 30 - 200 litres
- Temperatures from 0 to 60oC
- Models cooled by a hermetically sealed refrigeration system
- Internal heatproof toughened glass door allows clear view of inner chamber without disturbing the atmosphere
- High performance fan ensures optimum temperature uniformity
|
|
CO2 Laboratory Incubators
For professionals who need to simulate natural cellular growth environment |
|
|
Binder Air-Jacketd CO2 Incubators
- Capacities of 150 litres and 210 litres
- Temperature range of ambient +7oC to 100oC
- Precise temperature and CO2 control eliminates harmful overshoot
- Seamless stainless steel interior minimizes contamination risk
- Units features intelligent electronic control of temperature and infrared CO2 measurement
- Air jacket system ensures temperature accuracy and repeatable results
|