log in or register
You are filtering by:
Category:
Clear Category Laboratory Equipment
Clear Category Mixers, Blenders, and Stirrers
Clear Category Static (In-line) Mixers
Clear Category In-Line Static Mixers
Clear all filters
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
More Unique Lab & Industrial Supplies
In-Line Static Mixers
In-Line Static Mixers
Cole-Parmer offers a wide selection of in-line static mixers for use in flexible tubing and piping. Our range includes lengths up to 88 cm, materials including PVC, 316 Stainless Steel and PTFE and internal dimensions up to 5cm. To find the right static mixer for your application use Cole Parmer UK's on-line search or alternatively contact our Application Specialists for free technical support and let us bring over 50 years of mixing experience to help solve your application needs.
Static Mixers let you mix fluids as you pump them through a pipe line. The fluid moves through the mixer in an alternating clockwise/counterclockwise motion to ensure homogeneous product. They are virtually maintenance free and need no spare parts. A in line static mixer is efficient and can withstand pressures up to 100 bar with the smallest sizes. The mixing elements inside the pipe and tube mixers are securely attached to the walls of the mixer so that internal pressure changes cannot collapse elements and stop flow.
Unmounted mixer
Disposable In-line Static Mixers
  • Ideal for your one-time mixing applications.
  • These disposable polyacetal mixers are available unmounted or mounted in polypropylene tubing. Adjust unmounted mixer lengths to get optimal mixing action - shorten by cutting or extend by using more than one mixer.
  • An unmounted static mixer can be used with any standard tubing which has the same ID as the mixing element OD.
  • A mounted static mixer can be used with any compression fitting with the same tubing OD.
Clear PVC schedule 40 pipe mixer
Pipe In-line Static Mixers
  • Install this static mixer directly into your process line to achieve a 100% mixed output - mixing elements are welded inside the pipe.
  • All mixers have threaded ends with a choice of NPT connections up to 2"
  • Available in materials including PVC, PTFE and stainless steel
  • Clear PVC mixers to allow visual checks of your mixing process.
Sanitary Mixer, 1/2'
Sanitary In line Static Mixers
  • These are made of 316 stainless steel for low-or high-viscosity food processing.
  • All internal surfaces are ground and polished to a sanitary (#4) finish and have removable elements for cleaning and visual inspection.
  • Our blade design are best for low viscosity (<1,500 cp) fluids. The faster the liquid moves, the faster (and more thorough) the mixing. Great for mixing products into water or water-like fluids.
  • Our helical design models are best for high viscosities (>1500 Cp) fluids and when fluids are moving more slowly through the mixer. Excellent for combining products into fluids such as yogurt, mayonnaise and ice cream.
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
 Product Detail
Sort
 Availability
 Price
0466950.jpg - 316 SS tube mixer; 1/4" OD; 21 elements
316 SS tube mixer; 1/4" OD; 21 elements
Product Type Mixer,Brand KoFlo
WZ-04669-56
In stock.
$132.00 USD/EACH
Qty:
0466881.jpg - Sanitary Mixer, helical design, 2
Sanitary Mixer, helical design, 2
Product Type Mixer,Clamp size 2 in Tri-Clamp
WZ-04668-84
Usually ships in 21 days.
$5,072.00 USD/EACH
Qty:
0466702.jpg - Unmounted in-line mixer; 3/8"OD, 12 elements, 10/pack
WZ-04667-06
In stock.
$40.00 USD/PACKAGE of 10
Qty:
0466985.jpg - Clear PVC schedule 40 pipe mixer; 3/4" NPT(M), 12 elements
WZ-04669-93
In stock.
$119.00 USD/EACH
Qty:
0466985.jpg - Clear PVC schedule 40 pipe mixer; 1/2" NPT(M), 12 elements
WZ-04669-91
In stock.
$108.00 USD/EACH
Qty:
0466866.jpg - PVC Mixer with Injection Port, 4" Line size
PVC Mixer with Injection Port, 4" Line size
Product Type Mixer,Flow range 90 to 250 GPM,Brand KoFlo
WZ-04668-70
Usually ships in 7 days.
$1,019.00 USD/EACH
Qty:
04669-12 image shown as example only.  See product description for individual details.
Pipe Mixer, 316 stainless steel, 1" NPT(M), 6 elements
Product Type Mixer,Material Stainless steel,Brand KoFlo
WZ-04669-16
In stock.
$244.00 USD/EACH
Qty:
0466985.jpg - Clear PVC schedule 40 pipe mixer; 1" NPT(M), 6 elements
WZ-04669-94
In stock.
$114.00 USD/EACH
Qty:
0466950.jpg - 316 SS tube mixer; 1/4" OD; 27 elements
316 SS tube mixer; 1/4" OD; 27 elements
Product Type Mixer,Brand KoFlo
WZ-04669-58
In stock.
$145.00 USD/EACH
Qty:
0466866.jpg - PVC Mixer with Injection Port, 2-1/2" Line size
PVC Mixer with Injection Port, 2-1/2" Line size
Product Type Mixer,Flow range 35 to 100 GPM,Brand KoFlo
WZ-04668-66
Usually ships in 7 days.
$667.00 USD/EACH
Qty:
0466985.jpg - Clear PVC schedule 40 pipe mixer; 2" NPT(M), 6 elements
WZ-04669-98
Usually ships in 4 days.
$166.00 USD/EACH
Qty:
0466844.jpg - Sanitary Mixer, 1 1/2" Tri-clamp size, 6 elements
Sanitary Mixer, 1 1/2" Tri-clamp size, 6 elements
Product Type Mixer,Clamp size 1.5 in Tri-Clamp
WZ-04668-40
Usually ships in 14 days.
$2,457.00 USD/EACH
Qty:
04669-12 image shown
Pipe Mixer, 316 stainless steel, 1/2" NPT(M), 6 elements
Product Type Mixer,Material Stainless steel,Brand KoFlo
WZ-04669-12
In stock.
$224.00 USD/EACH
Qty:
0466974.jpg - PVC static mixer without injection port; 4" dia
PVC static mixer without injection port; 4" dia
Product Type Mixer,Flow range 90 to 250 GPM,Brand KoFlo
WZ-04668-71
Usually ships in 8 days.
$834.00 USD/EACH
Qty:
0466950.jpg - 316 SS tube mixer; 1/4" OD; 34 elements
316 SS tube mixer; 1/4" OD; 34 elements
Product Type Mixer,Brand KoFlo
WZ-04669-60
In stock.
$159.00 USD/EACH
Qty:
116 results (showing 1 - 15 )
Per Page