Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Price Range:
Capacity (gal.):
1-5 (1)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
 
Home  >  Document and Waste Containers

Document and Waste Containers

Document and Waste Containers

A stainless steel waste bin is an excellent addition to any lab especially one dealing with waste produced in chemical applications. Though proper waste storage protocols must be handled for hazardous waste, a stainless steel waste bin is perfect for other types of wastes that needs disposal. Cole-Parmer has a selection that showcase a stainless steel waste bin product which doesn’t rust or erode or even carry a charge. You can also find a stainless steel waste bin that lives up to OBRA regulations and OSHA standards on bloodborne pathogens and the safe containment of infectious waste. The aforementioned stainless steel waste bin is both UL rated and FM approved adding an extra layer of reassurance for your research facility. There are other trash containers including those made form polymer or a steel plate or stainless steel. The polymer and steel plate versions comply with HIPAA regulations for use in situations where organizations are liable for protecting patient or employee privacy. For help on selecting an applicable stainless steel waste bin or other trash receptacle contact a Technical Expert today.
 Product Detail
Sort
 Availability
 Price

Defenders Stainless Steel Receptacle, 24 gallon

  • Product Type: Waste Bins
  • Dimensions: 15 in x 15 in x 30 in
  • Dimensions: 15 " W
WZ-47580-12
Available for order
or call 1-888-358-4705 to order
$775.00 USD/EACH

Qty:

Defenders Stainless Steel Receptacle, 7 gallon

  • Product Type: Waste Bins
  • Dimensions: 12 in x 12 in x 17 in
  • Dimensions: 12 " W
WZ-47580-08
Available for order
or call 1-888-358-4705 to order
$573.00 USD/EACH

Qty:

Defenders Stainless Steel Receptacle, 3 1/2 gallon

  • Product Type: Waste Bins
  • Dimensions: 11 in Dia. x 17 in
  • Dimensions: 17 " H
WZ-47580-06
Usually ships in 15 days.
$289.00 USD/EACH

Qty:

3 results (showing 1 - 3 )
Per Page 
#PRODUCTTITLE#

#PRODUCTHEADER#

#IAMGESTICKER#
#CROSSOUTPRICE#
#PRICE#
#PRICEWITHVAT#
#AVAILABILITY#
Qty
Quantity Discounts
Qty: #QTY10COUNT# #QTY15COUNT# #QTY20COUNT#
Price: #QTY10PRICE# #QTY15PRICE# #QTY20PRICE#
#PRODUCTIMAGE#
#POWERREVIEW#
#MARKETINGTEXT#
#ICONS#
#SPECS#
Home  >  Document and Waste Containers