Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Dissolved Oxygen Benchtop Meters
Dissolved Oxygen Benchtop Meters
Getting a dissolved oxygen measurement can be tricky. A stable and accurate dissolved oxygen measurement reading is critical. Yet, dissolved oxygen can be affected by water temperature, salinity, and barometric pressure. Factors such as exposure to dirty or wet environments can be disrupting when trying to secure an accurate dissolved oxygen measurement. To minimize contamination of dissolved oxygen measurement researchers often use benchtop oxygen in a controlled laboratory setting. General research or water quality testing laboratories are good examples of applications using benchtop dissolved oxygen meters. Wastewater and environmental facilities utilize these meters to measure a derivation of dissolved oxygen known as biological oxygen demand (BOD). This EPA-required determination of bacterial activity in water samples requires the controlled conditions and accuracy of a laboratory setting. There are a host of such meters available from industry-leading brands such as YSI, Oakton, Cole-Parmer, Thermo Scientific Orion, accumet, Hach, and others. For help in selecting the right tool to ensure an accurate dissolved oxygen measurement contact a Technical Expert today.
 Product Detail
Sort
 Availability
 Price
WZ-35415-00 Oakton<small><sup>&reg;</sup></small> DO 700 Benchtop Dissolved Oxygen Meter
Oakton DO 700 Benchtop Dissolved Oxygen Meter
Product Type Dissolved Oxygen Meter,Meter style Benchtop,Dimensions " W 6-7/8
WZ-35415-00
In stock.
$995.00 USD/EACH
Qty:
Representative photo only YSI Model 5100 Benchtop Dissolved Oxygen Meter 115 VAC
YSI Model 5100 Benchtop Dissolved Oxygen Meter, 115 VAC
Product Type Dissolved Oxygen Meter,Meter style Benchtop,Dimensions " W 9
WZ-05510-22
Usually ships in 15 days.
$1,802.30 USD/EACH
Qty:
Representative photo only Oakton DO 700 Benchtop Dissolved Oxygen Meter with NIST traceable Calibration
Oakton DO 700 Benchtop Dissolved Oxygen Meter with NIST-traceable Calibration
Product Type Dissolved Oxygen Meter,Meter style Benchtop,Dimensions " W 6-7/8
WZ-35415-01
Call 1-888-358-4705 for availability.
$1,098.00 USD/EACH
Qty:
WZ-53101-06 THERMO SCIENTIFIC ORION STAR A213 DO/BOD BENCHTOP METER KIT
Thermo Scientific Orion Star A213 DO/BOD Benchtop Meter Kit
Product Type Dissolved Oxygen Meter,Meter style Portable,Dimensions " W 7
WZ-53101-06
Usually ships in 6 days.
$2,225.00 USD/EACH
Qty:
Representative photo only Oakton DO 2700 Benchtop Dissolved Oxygen Meter
Oakton DO 2700 Benchtop Dissolved Oxygen Meter
Product Type Dissolved Oxygen Meter,Meter style Benchtop,Dimensions " W 6-7/8
WZ-35416-00
In stock.
$1,900.50 USD/EACH
Qty:
WZ-53101-16 THERMO SCIENTIFIC ORION STAR A223 DO PORTABLE METER
Thermo Scientific Orion Star A223 DO Portable Meter Only
Meter style Portable,Temperature compensation Automatic
WZ-53101-16
Usually ships in 6 days.
$762.00 USD/EACH
Qty:
WZ-99561-24 OpTech<small><sup>&reg;</sup></small> Rapid BOD/CBOD Analyzer
OpTech Rapid BOD/CBOD Analyzer
Product Type Dissolved Oxygen Meter,Meter style Benchtop,Dimensions " W 1 1/2
WZ-99561-24
Usually ships in 8 days.
$7,995.00 USD/EACH
Qty:
WZ-59004-24 HACH HQD DIGITAL PH/CON/DO BENCH KIT
Hach HQ440d Digital pH/Con/LDO Benchtop Meter Kit
Product Type Multiparameter Meter,Meter style Benchtop,Dimensions " W 6 7/8
WZ-59004-24
Usually ships in 4 days.
$3,290.00 USD/EACH
Qty:
WZ-53105-24 INOLAB 7310 ADVANCED DO BENCHTOP METER WITH PRINTER
inoLab 7310 advanced DO benchtop meter with printer
Product Type Dissolved Oxygen Meter,Meter style Benchtop,Dimensions " W 7
WZ-53105-24
Usually ships in 6 days.
$2,203.00 USD/EACH
Qty:
WZ-05510-08 Multilab Meter
YSI Multilab Advanced BOD/pH Benchtop Lab Kit
Product Type Dissolved Oxygen Meter,Meter style Benchtop,Dimensions " W 7.5
WZ-05510-08
Usually ships in 10 days.
$3,000.00 USD/EACH
Qty:
WZ-53002-21 Strathkelvin Precision Dissolved Oxygen Respirometer, six-channel
WZ-53002-21
Call 1-888-358-4705 for availability.
$10,050.00 USD/EACH
Qty:
WZ-05510-25 Image may vary from actual product
YSI Model 5000 Benchtop Dissolved Oxygen Meter, 230 VAC
Product Type Dissolved Oxygen Meter,Meter style Benchtop,Dimensions " W 9
WZ-05510-25
Call 1-888-358-4705 for availability.
$1,477.35 USD/EACH
Qty:
WZ-99561-57 REFCHECK STANDARD REFERENCE VIALS - 15.0ML
WZ-99561-57
Usually ships in 8 days.
$765.00 USD/PACKAGE
Qty:
WZ-53105-20 INOLAB 7310 ADVANCED DO BENCHTOP METER WITH GALVANIC PROBE
inoLab 7310 advanced DO benchtop meter with galvanic probe
Product Type Dissolved Oxygen Meter,Meter style Benchtop,Dimensions " W 7
WZ-53105-20
Usually ships in 6 days.
$2,325.00 USD/EACH
Qty:
WZ-59004-26 HACH HQD DIGITAL BOD BENCH KIT
Hach HQd Digital BOD Bench Kit
Product Type Dissolved Oxygen Meter,Meter style Benchtop,Dimensions " W 6 7/8
WZ-59004-26
Usually ships in 4 days.
$2,725.00 USD/EACH
Qty:
35 results (showing 1 - 15 )
Per Page 
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View Categories By: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title