Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
You are filtering by:
Category:
Clear Category Pumps
Clear Category Metering Pumps
Clear Category Diaphragm Pumps
Clear all filters
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334

More Unique Lab & Industrial Supplies

 

Diaphragm Pumps

Diaphragm Pumps
Diaphragm pump technology uses the force of a flexible diaphragm to displace liquid with each stroke. Using this principle, Cole-Parmer's range of diaphragm pumps are able to transfer liquids with a flow rate of 0.07 mL/min to 560 LPM and pressures up to 30 bar.

Cole-Parmer offer three types of diaphragm pump technology with varying characteristics:

Digital remote contr

Solenoid Diaphragm Pump

  • Flow rates - 0.08 mL/min to 250 LPH
  • Pressures up to 17.3 bar
  • Use for controlled metering and dosing applications

Solenoid diaphragm pumps are known as metering pumps as they provide precise control of flow rates delivering from 0.08 ml/min up to 4,167 ml/min with pressures up to 17.3 bar. Our range includes manual and digital adjustable stroke lengths and stroke frequency to enable flow control. Pump models are available with external triggers and input control options and come in a variety of materials for the internals parts as well as the housing. These pumps requires no seals so you can pump abrasives and slurries if the valves are kept clean

Moderate Flow Positi

Positive Displacement Diaphragm Pump

  • Flow rates to 64 LPM
  • Pressures up to 30 bar
  • Use for general transfer and spraying applications

These types of diaphragm pumps are known as positive displacement pumps as they deliver a definite volume of liquid using one or two flexible diaphragms for each cycle of pump operation. Ideal for general transfer and spraying applications, our range can offer flow rates of up to 64 LPM, pressures up to 30 bar and with material options to allow compatibility with corrosive liquids. Many of our diaphragm pumps include factory set pressure limits to either shut the pump off or open a by-pass circuit within the pump to protect the pump and system from damage.

High-flow, air-drive

Air Operated Diaphragm Pump

  • Flow rates to 560 LPM
  • Pressures up to 8.3 bar
  • Use for general transfer applications and are ideal for transferring viscous and slurry media.

Air operated diaphragm pumps are often referred to as a AOD pumps or AODD pumps. They use the same technology as positive displacement diaphragm pumps however they contain no heat generating electrical components allowing safe, explosion-proof operation in hazardous areas. Used extensively in the food and paints industry for general transfer applications they are able to deliver flow rates up to 560 LPM. AOD pumps have the ability to handle low to high viscosity fluids and even particulates. They have the ability to self prime and are available in a variety of materials including PTFE and stainless steel.

Use Cole-Parmer's on-line catalogue to find and compare pump specifications. Alternatively contact our Application Specialists for free technical support and let us bring over 50 years of pumping experience to help solve your application needs.

Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
 Product Detail
Sort
 Availability
 Price
PTFE-Diaphragm Pump Head

PTFE-diaphragm pump head, 80 to 800 mL/min

Product Type Diaphragm Metering Pumps,Wetted parts PTFE diaphragm, body, valve seat, spring, ball check,Max pressure 50 psi
WZ-07090-42
In stock.
$594.00 USD/EACH

Qty:

PTFE-Diaphragm Pump Head

PTFE-diaphragm pump head, 10 to 100 mL/min

Product Type Diaphragm Metering Pumps,Wetted parts PTFE diaphragm, body, valve seat, spring; borosilicate glass ball check,Max pressure 75 psi
WZ-07090-62
In stock.
$604.00 USD/EACH

Qty:

7414010.jpg - Manual control pump, 44 gdp, 115 VAC

Manual control pump, 44 gdp, 115 VAC

Product Type Diaphragm Metering Pumps,Wetted parts PVC, PTFE, ceramic,Max pressure 100 psi
WZ-74142-00
In stock.
$643.00 USD/EACH

Qty:

74148_05.jpg - Pump, Metering, 65gph, 115/230V, Stainless Steel

Pump, Metering, 65gph, 115/230V, Stainless Steel

Product Type Diaphragm Metering Pumps,Max pressure 150 psi,Max flow rate mL/min 4100
WZ-74148-70
Usually ships in 30 days.
$2,591.00 USD/EACH

Qty:

7450740.jpg - Remotel control solenoid diaphragm metering pump, 8.0 GPH, 115 VAC

Remotel control solenoid diaphragm metering pump, 8.0 GPH, 115 VAC

Product Type Diaphragm Metering Pumps,Wetted parts PVC head and fittings, ceramic balls, Fluorofilm diaphragm, PVDF/Polyprel seal ring,Max pressure 60 psi
WZ-74514-40
Usually ships in 11 days.
$1,461.00 USD/EACH

Qty:

7414010.jpg - 4 to 20 mA pace pumps, 42 GPD, 230 VAC

4 to 20 mA pace pumps, 42 GPD, 230 VAC

Product Type Diaphragm Metering Pumps,Wetted parts PVC, PTFE, ceramic,Max pressure 150 psi
WZ-74141-35
Usually ships in 18 days.
$1,256.00 USD/EACH

Qty:

7630240.jpg - High-volume, single head Cole-Parmer<reg> chemical feed solenoid diaphragm pump, 662 mL/min, 115 VAC

High-volume, single head Cole-Parmer chemical feed solenoid diaphragm pump, 662 mL/min, 115 VAC

Product Type Diaphragm Metering Pumps,Wetted parts polypropylene pump head, PVDF cartridge fittings, ceramic ball valves, HASTELLOY C® springs, O-rings of VITON® fluoroelastomer, ethylene propylene (EP) diaphragm coated with TFE,Max pressure 10 psi
WZ-76302-40
In stock.
$438.00 USD/EACH

Qty:

7452100.jpg - Digital remote control solenoid diaphragm metering pump, 1.0 GPH, 115 VAC

Digital remote control solenoid diaphragm metering pump, 1.0 GPH, 115 VAC

Product Type Diaphragm Metering Pumps,Wetted parts PVC head and fittings, ceramic balls, Fluorofilm diaphragm, PVDF/Polyprel seal ring,Max pressure 110 psi
WZ-74521-10
Usually ships in 9 days.
$1,290.00 USD/EACH

Qty:

7450740.jpg - Remotel control solenoid diaphragm metering pump, 4.0 GPH, 115 VAC

Remotel control solenoid diaphragm metering pump, 4.0 GPH, 115 VAC

Product Type Diaphragm Metering Pumps,Wetted parts PVC head and fittings, ceramic balls, Fluorofilm diaphragm, PVDF/Polyprel seal ring,Max pressure 100 psi
WZ-74514-30
In stock.
$1,367.00 USD/EACH

Qty:

7414010.jpg - Manual control pump, 94 gdp, 115 VAC

Manual control pump, 94 gdp, 115 VAC

Product Type Diaphragm Metering Pumps,Wetted parts PVC, PTFE, ceramic,Max pressure 100 psi
WZ-74142-10
Usually ships in 11 days.
$908.00 USD/EACH

Qty:

7450315Back.jpg - Manual control solenoid diaphragm metering pump, 2.5 GPH, 115 VAC

Manual control solenoid diaphragm metering pump, 2.5 GPH, 115 VAC

Product Type Diaphragm Metering Pumps,Wetted parts PVC head and fittings, ceramic balls, Fluorofilm diaphragm, PVDF/Polyprel seal ring,Max pressure 100 psi
WZ-74513-20
Usually ships in 25 days.
$989.00 USD/EACH

Qty:

no_image.gif - Programmable Remote Control Solenoid-driven Diaphragm Metering Pump with GFRPP head; 8.5 GPH, 230 VAC

Programmable Remote Control Solenoid-driven Diaphragm Metering Pump with GFRPP head; 8.5 GPH, 230 VAC

Product Type Diaphragm Metering Pumps,Wetted parts Pump head and fittings: glass fiber reinforced polypropylene (GFRPP)
Diaphragm: PTFE bonded to EPDM
Ball check valves: alumina ceramic
Seals: fluoroelastomer
O-rings: PTFE,Max pressure 105 psi
WZ-74125-15
Usually ships in 58 days.
$1,170.00 USD/EACH

Qty:

7414130.jpg - High-viscosity, 4 to 20 mA pace pump, 24 GPD, 115 VAC, 50/60 Hz

High-viscosity, 4 to 20 mA pace pump, 24 GPD, 115 VAC, 50/60 Hz

Product Type Diaphragm Metering Pumps,Wetted parts PVC, PTFE, 316 SS,Max pressure 150 psi
WZ-74147-10
Usually ships in 10 days.
$1,165.00 USD/EACH

Qty:

no_image.gif - Programmable Remote Control Solenoid-driven Diaphragm Metering Pump with GFRPP head; 5.5 GPH, 115 VAC

Programmable Remote Control Solenoid-driven Diaphragm Metering Pump with GFRPP head; 5.5 GPH, 115 VAC

Product Type Diaphragm Metering Pumps,Wetted parts Pump head and fittings: glass fiber reinforced polypropylene (GFRPP)
Diaphragm: PTFE bonded to EPDM
Ball check valves: alumina ceramic
Seals: fluoroelastomer
O-rings: PTFE,Max pressure 150 psi
WZ-74125-00
Usually ships in 14 days.
$1,117.00 USD/EACH

Qty:

7380300.jpg - Manual control feed verification diaphragm pump, 11.4 GPD, PVDF, 94 to 264 VAC

Manual control feed verification diaphragm pump, 11.4 GPD, PVDF, 94 to 264 VAC

Product Type Diaphragm Metering Pumps,Wetted parts Pump head, foot valve/strainer: PVDF; Diaphragm: PTFE; Valve seat: Viton; Check ball: Ceramic,Max pressure 140 psi
WZ-73803-40
Usually ships in 4 days.
$404.00 USD/EACH

Qty:

230 results (showing 1 - 15 )
Per Page