Cole-Parmer - The world's leading source for scientific and industrial instrumentation since 1955.
United Kingdom   Cart  Subtotal = 0.00
0500 345 300 Items = 0  View Cart
Login Login
New Customer? Register
Home Products & Services Technical Library myCP Account Contact/About Us
Cole-Parmer UK Catalog  > Laboratory Equipment  > Balances 
Balances, Weighing Scales, Weighing Products
  More Information Buy
Baths Baths & Circulators Shop online
Centrifuges" Centrifuges Shop online
Furnaces" Furnaces Shop online
Refrigerators Refrigerators & Freezers Shop online
Ultrasonic Ultrasonic Cleaners Shop online
Cole-Parmer UK is a leading distributor of weighing products. Whether you are looking for laboratory balances, laboratory scales or industrial scales, we are able to offer a wide selection for all your needs. Our range contains leading brands including – Cole-Parmer Symmetry, Ohaus, Mettler Toledo, A&D Engineering, Sartorius, Acculab and Adam Equipment. Our full range of balances can either be viewed online or from our general catalogue which contains over 40 pages of laboratory balances and scales.
Cole-Parmer’s balance range includes :
Acculab VICON <small><sup>®</sup></small>  P
Portable and pocket balances
  • Capacities up to 12,000g
  • Readability of up to 0.001g
210g, 120VAC
Toploading balances
  • Capacities up to 16,000g
  • Readability of up to 0.001g
Analytical Balances,
Analytical balances
  • Capacities up to 320g
  • Readability of up to 0.01mg
2.1g, 115VAC, Microb
Microbalances
  • Capacities up to 610g
  • Readability of up to 0.1ug
Ohaus MB 45 Advanced
Moisture balances
  • Capacities up to 81g
  • Readability of up to 0.01%
Acculab SVI-Series B
Industrial scales
  • Capacities up to 300kg
  • Readability of up to 0.0005 kg
balance1
Calibration masses
  • Masses up to 20kg in size
  • NVLAP and UKAS certification available
balance2
Weighing accessories
  • Includes balance draft shields, printing supplies and weighing dishes
Our range in laboratory balances and industrial scales includes a variety of features to suit your application needs. Models are available with internal or external calibration, built in RS-232 to allow connectivity to computers or printers and stability filters to compensate from vibrations, drafts or temperature changes. To find the right laboratory balance and industrial scales for your application use Cole-Parmer’s on-line search to help you shop by specification or by brand. Alternatively contact our Application Specialists for free technical support and let us bring over 50 years of weighing experience to help solve your application needs.
Privacy Statement  |  Terms and Conditions  |  Careers@Cole-Parmer  |  Press Releases  |  Site Map
Cole-Parmer Sites
Cole-Parmer: United States
  Canada
  China
  United Kingdom
  India
MasterFlex.com - Ordering and Technical Info
FoodTechSource.com - Food Quality Site
InnocalSolutions.com - Calibration and Certification
Cole-Parmer
Unit 3 River Brent Business Park
Trumpers Way, Hanwell, London W7 2QA
Telephone: 020 8574 7556
Fax: 020 8574 7543
E-Mail: sales@Coleparmer.co.uk
Copyright © 2007